Learn More
Sartorius Quintix™ Pro Motorized Leveling Laboratory Balance
GREENER_CHOICE
Quintix™ Pro laboratory balances are not just about meeting your expectations but exceeding them. They come with an array of premium features that enhance usability, flexibility, and incorporate an eco-friendly design.
Brand:  Sartorius QTX3202IMO-1S
 View more versions of this product
Product Code. 18002184
  • 4410.00 EUR / Each
Explore more special offers
Description
Description
  • Enhance the user experience with new premium features.
  • Offering a wide range of use in different environments and applications.
  • Energy efficiency, reduced carbon footprint, and sustainability, minimizing environmental impact from the manufacturing plant to the bench.
  • Real-time level
TRUSTED_SUSTAINABILITY
Specifications
Specifications
D-sub Connector, 9-pin
Touchscreen
Standard Balance Without Country-specific Additions
Quintix Pro Balance
Balance
No
3200 g
10 mg
Motorized
≤ 0.9 s
100 to 240 VAC (± 10%) 50 to 60 Hz 0.2 A