missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2,2-Dimethyl-1,3-dioxolane-4-methanol, 98%, Thermo Scientific Chemicals

Product Code. 10390594 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10390594

Brand: Thermo Scientific Acros 221330050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14347-78-5
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00003213
InChI Key RNVYQYLELCKWAN-RXMQYKEDSA-N
Synonym r---2,2-dimethyl-1,3-dioxolane-4-methanol, r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, 4r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, r-solketal, r---1,2-o-isopropylideneglycerol, r-2,2-dimethyl-1,3-dioxolane-4-methanol, 2,3-o-isopropylidene-sn-glycerol, d-acetone glycerol, l---1,2-isopropylideneglycerol, r---2,3-o-isopropylideneglycerol
PubChem CID 736056
IUPAC Name [(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
SMILES CC1(OCC(O1)CO)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 72.0°C to 73.0°C (8.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4330 to 1.4350
Quantity 5 g
Beilstein 19, III, 646
Merck Index 10, 3236
Specific Gravity 1.06
Specific Rotation Condition - 14.00 (20.00°C neat)
Specific Rotation − 14.00
Solubility Information Solubility in water: miscible
Formula Weight 132.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (R)-(-)-2, 2-Dimethyl-1, 3-dioxolane-4-methanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.