missing translation for 'onlineSavingsMsg'
Learn More

R-268712, MedChemExpress

Product Code. 16478157
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16478157

Brand: Medchem Express HY12953

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress R-268712 is an orally active and selective ALK-5 inhibitor, with an IC50 of 2.5 nM. R-268712 inhibits the phosphorylation of Smad3 in a dose-dependent manner with an IC50 of 10.4 nM. R-268712 suppresses glomerulonephritis as well as glomerulosclerosis by inhibiting TGF-β signaling, which can be used in studies of renal fibrosis and cancer.

R-268712 is an orally active and selective ALK-5 inhibitor, with an IC50 of 2.5 nM. R-268712 inhibits the phosphorylation of Smad3 in a dose-dependent manner with an IC50 of 10.4 nM. R-268712 suppresses glomerulonephritis as well as glomerulosclerosis by inhibiting TGF-β signaling, which can be used in studies of renal fibrosis and cancer.

Specifications

Chemical Name or Material R-268712
CAS 879487-87-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H18FN5O
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (343.98 mM; Need ultrasonic)
SMILES FC1=C(C2=CN(CCO)N=C2)C=C(C3=CNN=C3C4=CC=CC(C)=N4)C=C1
Molecular Weight (g/mol) 363.39
Formula Weight 363.39
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.