Learn More
R&D Systems™ Mouse NKG2D/CD314 (NP_149069) VersaClone cDNA
Natural Killer G2D
Brand: R&D Systems™ RDC0851
This item is not returnable.
View return policy
Description
The Mouse NKG2D/CD314 (NP_149069) VersaClone cDNA clone contains the complete ORF for NKG2D/CD314, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The NKG2D/CD314 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
mNKG2D | |
10 μg at 0.2 μg/μL | |
712bp | |
Mouse |
NP_149069 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans