Learn More
R&D Systems™ Rat CXCR5 (NP_445755) VersaClone cDNA
Chemokine (C-X-C motif) Receptor 5
Brand:  R&D Systems™ RDC0037
Catalog No. 18441710
  • 322.00 EUR / 10µg
Description
The Rat CXCR5 (NP_445755) VersaClone cDNA clone contains the complete ORF for CXCR5, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CXCR5 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
rCXCR5
10 μg at 0.2 μg/μL
1138bp
Rat
NP_445755
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Certificates
Promotions
Promotions
For Reasearch Use Only; Not for Use in Humans