Learn More
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine, 97%, Thermo Scientific Chemicals
Description
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine acts as a ligand used in the enantioselective vinylation of aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine acts as a ligand used in the enantioselective vinylation of aldehydes.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 141°C to 143°C |
| Quantity | 500 mg |
| Optical Rotation | −355° (c=0.6 in Ethanol) |
| Formula Weight | 441.34 |
| Percent Purity | 97% |
| Chemical Name or Material | (R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.