Learn More
(R,R)-BD-AcAc 2, MedChemExpress
MedChemExpress (R,R)-BD-AcAc 2 ((R,R)-Ketone Ester), a ketone monoester, can be used as a source of oral nutritional ketones.
Brand: Medchem Express HY-15344
This item is not returnable.
View return policy
Description
(R,R)-BD-AcAc 2 can elevate plasma levels of β-hydroxybutyrate and acetoacetate, blood glucose, blood Na+ levels and blood creatinine levels after oral administration in mice. (R,R)-BD-AcAc 2 can partly prevent muscle weakness in septic mice. (R,R)-BD-AcAc 2 has potential to improve exercise performance and endurance in animal body. (R,R)-BD-AcAc 2 can also be used to research Parkinson’s disease or diabetes.Specifications
(R,R)-BD-AcAc 2 | |
1208313-97-6 | |
500 mg | |
C8H16O4 | |
(R,R)-Ketone Ester | |
C[C@@H](O)CC(OCC[C@H](O)C)=O | |
176.21 | |
Research |
Viscous Liquid | |
Light Yellow | |
Research | |
Neuroscience-Neurodegeneration | |
H2O : 100 mg/mL (567.50 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (567.50 mM; Need ultrasonic) | |
176.21 | |
95.0% |