Learn More
Epredia™ RA Lamb Biopsy Inserts, Biopsy insert pad, white
Prevent the loss of small or friable material while allowing interchange of fluids with Epredia™ RA Lamb Biopsy Inserts.
58.65 EUR
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
12628366
|
Epredia™
E10/87/B |
Insert Pad | N/A | ||||||
Description
- Mesh fits inside processing cassettes
- Mesh hole size is 0.5mm
Specifications
Stainless Steel |