Learn More
Rabbit anti-Human IgG Fc Secondary Antibody, AP, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen 31318
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31318 has been successfully used in Western blot and IP applications. Product # 31318 reacts with the heavy chains of human IgG, but not with the light chains common to most human immunoglobulins. The antibody does not react against human IgM, IgA, or against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 1.0 mL of distilled water (0.6 mg/mL after restoration). Country of Origin: USA
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Human IgG Fc | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Rabbit | |
1 mL | |
Secondary | |
4° C | |
IgG |
Western Blot | |
0.6 mg/mL | |
TBS with 15mg/mL BSA and 0.05% sodium azide; pH 8 | |
Antigen affinity chromatography | |
RUO | |
Human | |
Lyophilized |