Learn More
Rabbit anti-Mouse IgA Fc Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69205
Product Code. 18681758
  • 711.00 EUR / 10mg
Explore more special offers
Description
Description
IgA Fc Polyclonal antibody specifically detects IgA Fc in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoblot, Dot Blot
Specifications
Specifications
IgA Fc
Polyclonal
Unconjugated
Lyophilized from PBS (pH 7.2)
Purified homogenous IgA isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
10 mg
Secondary
Mouse
Lyophilized
Western Blot, ELISA, Immunofluorescence, Immunoblot, Dot Blot
LYOPH
Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000
Rabbit
Ion exchange chromatography
RUO
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.