missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG1 Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-69521
This item is not returnable.
View return policy
Description
IgG1 Polyclonal antibody specifically detects IgG1 in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoblot, Dot BlotSpecifications
IgG1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from PBS (pH 7.2) | |
Rabbit | |
Ion exchange chromatography | |
RUO | |
Secondary | |
Mouse | |
Lyophilized |
Western Blot, ELISA, Immunofluorescence, Immunoblot, Dot Blot | |
LYOPH | |
Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000 | |
IgG1, Igh-4, RGD1359539, VH7183 | |
Pools of highly purified homogenous IgG1 isolated from mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
10 mg | |
Immunology | |
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction