Learn More
Rabbit anti-Mouse IgG2b Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-69599
Product Code. 18658958
  • 810.00 EUR / 10mg
Explore more special offers
Description
Description
IgG2b Polyclonal antibody specifically detects IgG2b in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoblot, Dot Blot
Specifications
Specifications
IgG2b
Polyclonal
Unconjugated
Lyophilized from PBS (pH 7.2)
Pools of purified homogenous IgG2b isolated from pooled mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
10 mg
Neuronal Cell Markers, Neuroscience, Neurotransmission
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot, ELISA, Immunofluorescence, Immunoblot, Dot Blot
LYOPH
Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000
Rabbit
Ion exchange chromatography
RUO
Secondary
Mouse
Lyophilized