missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG3 Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-69632
This item is not returnable.
View return policy
Description
IgG3 Polyclonal antibody specifically detects IgG3 in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoblot, Dot BlotSpecifications
IgG3 | |
Polyclonal | |
Lyophilized from PBS (pH 7.2) | |
Rabbit | |
Ion exchange chromatography | |
RUO | |
Secondary | |
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunofluorescence, Immunoblot, Dot Blot | |
Unconjugated | |
DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809 | |
Pools of purified homogenous IgG3 isolated from pooled mouse serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
10 mg | |
Immunology | |
3502 | |
Mouse | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction