Learn More
MP Biomedicals Rabbit Mineral Mix
Rabbit diet with minerals.
Brand:  MP Biomedicals 0296026302
 View more versions of this product
Product Code. 11437191
  • 96.35 EUR / 2kg
Description
Provides complete minerals for normal growth and development of rabbit when added at a level of 6.0% of the diet.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Not Autoclavable
Single-use
Non-sterile
1 ft.
Animal Diet
2 kg
Room Temperature
0.3048 m
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.