missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Rabbit Serum

Sterile, Frozen Brown Solution. Combine with BSA to use as a blocking serum in immunocytochemistry and immunohistochemistry.

Brand:  MP Biomedicals™ 0219135720

 View more versions of this product

Product Code. 11470422

  • 43.67 EUR / 20mL
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

-20°C
Liquid
20 mL
Sterile
Blocking agent in immunohistochemistry culture media component
Rabbit
Rabbit Serum
4 to 7 g/dL
Membrane-filtered
6.5 to 8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers