Learn More
MP Biomedicals™ Rabbit Serum
Sterile, Frozen Brown Solution. Combine with BSA to use as a blocking serum in immunocytochemistry and immunohistochemistry.
Brand: MP Biomedicals™ 0219135780
Additional Details : Weight : 0.10000kg
Specifications
U.S. regulations | |
Rabbit | |
Rabbit Serum | |
4 to 7 g/dL | |
Membrane-filtered | |
6.5 to 8 |
-20°C | |
Liquid | |
100 mL | |
Sterile | |
Blocking agent in immunohistochemistry culture media component |