Learn More
Memmert™ Rack for shaking device for 119 test tubes
Product Code. 17140315 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
WTB15
WTB24
Unit Size:
Each
Product Code. 17140315
Brand: Memmert™ WTB15B48818
Currently Unavailable: We have no information when this product will be available.
View alternative products
Rack for shaking device for 90 test tubes
  • Rack for shaking device for test tubes
  • Accessory
Specifications
For Use With (Equipment) WTB15
Type Rack for shaking device for 119 test tubes 18 mm
Diameter (Metric) 18 mm