Learn More
Memmert™ Rack for shaking device for 45 test tubes
Product Code. 17150315 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
WTB15
WTB24
Unit Size:
Each
Product Code. 17150315
Brand: Memmert™ WTB15B48820
Only null left
Add to basket
Add to basket
Rack for shaking device for 25 test tubes
  • Rack for shaking device for test tubes
  • Accessory
Specifications
For Use With (Equipment) WTB15
Type Rack for shaking device for 45 test tubes 32 mm
Diameter (Metric) 32 mm