missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAIDD/CRADD Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-00051AF750
This item is not returnable.
View return policy
Description
RAIDD/CRADD Polyclonal specifically detects RAIDD/CRADD in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RAIDD/CRADD | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
CASP2 and RIPK1 domain containing adaptor with death domain, Caspase and RIP adapter with death domain, caspase and RIP adaptor with death domain, death domain containing protein CRADD, death domain-containing protein CRADD, MGC9163, RAIDDdeath adaptor molecule RAIDD, RIP-associated ICH1/CED3-homologous protein with death domain, RIP-associated protein with a death domain | |
Produced in rabbits immunized with purified, recombinant RAIDD/CRADD (P78560; Met1-Glu199). | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Tumor Suppressors | |
8738 | |
Store at 4°C in the dark. | |
IgG |