Learn More
Grant Instruments™ Raised Shelf for Unstirred Water Baths
Product Code. p-8014639
Click to view available options
Type:
Shelf for 12 L Tanks
Shelf for 18 L Tanks
Shelf for 26 L Tanks
Shelf for 34 L Tanks
Unit Size:
Each
Product Code. 11406487
Brand: Grant Instruments™ RS22
Accommodates shallow vessels. Grant Instruments™ Raised Shelf for Unstirred Water Baths can be used with a variety of other Grant Heating Circulators and Heated Circulating Baths.
  • Choose from a variety of sizes to fit your tank.
  • For use with either Stainless Steel and Plastic tank models
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Grant Heated Circulating Baths
Type Shelf for 18 L Tanks
Height (Metric) 40 or 135 mm