missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse IgM, Clone: II/41, Secondary Antibody, BD

Brand:  BD Biosciences 553435

214.89 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15857218

  • 215.00 EUR / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The II/41 clone has been reported to react specifically with mouse IgM of Igh-C[a] and Igh-C[b] haplotypes. It has been reported not to react with other Ig isotypes. In addition, the II/41 clone has been reported not to stimulate B-cell proliferation. Thi

Host Species: Rat
Clone: II/41
Isotype: IgG2a, κ
Species Reactivity: Mouse

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rat anti-Mouse IgM
II/41
Rat
0.5 mg
Immunology
Mouse
Monoclonal
0.5mg/mL
Affinity Purified
RUO
Secondary
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.