Learn More
B Braun™ RB Stainless Steel Needle
Brand:  B Braun™ 4665112
Additional Details : Weight : 0.12600kg
Product Code. 10296432
  • 7.35 EUR / Pack of 100
Estimated Shipment: 16-08-2024
Specifications
Stainless Steel
1.1 mm
Needle
Sterile
19 G
Yes
40 mm