Learn More
RBMX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17994-1-AP-20UL
 View more versions of this product
Product Code. 16898014
  • 140.00 EUR / 20µL
Explore more special offers
Specifications
RBMX2
Polyclonal
Unconjugated
RBMX2
CGI 79, RBMX2
Rabbit
Antigen Affinity Chromatography
RUO
209003, 367930, 51634
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
B0BN49, Q8R0F5, Q9Y388
Rbmx2
RBMX2 Fusion Protein Ag12439
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG