Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical, with Thermostatic Jacket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 06021273
 View more versions of this product
Product Code. 11572274
  • 1540.00 EUR / Each
Estimated Shipment: 13-01-2025
Description
  • Without graduation
  • With laboratory flange LF
  • With groove, with two connectors (with threads GL 14 /GL 18 or ball flanges KF 15)
  • Made of DURAN™ tubing
  • Other sizes on request
Specifications
Specifications
2 L
260 mm
1 Ea.
Duran Glass
Lab Flange 100 LF, Connector KF 15
206 mm