Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical, with Thermostatic Jacket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 06020279
This item is not returnable.
View return policy
Description
- Without graduation
- With laboratory flange LF
- With groove, with two connectors (with threads GL 14 /GL 18 or ball flanges KF 15)
- Made of DURAN™ tubing
- Other sizes on request
Specifications
4 L | |
295 mm | |
1 Ea. |
Duran Glass | |
Lab Flange 100 LF, Connector GL 18 | |
236 mm |