missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ ReagentPack™ Subculture Kit
Used for the passaging of primary cell types. Lonza™ ReagentPack™ Subculture Kits have been designed specifically for this purpose.
Brand: Lonza™ CC-5034
This item is not returnable.
View return policy
Description
Each kit contains 100 ml each of:- Trypsin/EDTA
- Trypsin Neutralizing Solution
- HEPES Buffered Saline
Specifications
ReagentPack containing: Trypsin/EDTA, 100mL, HEPES Buffered Saline Solution, 100mL Trypsin Neutralizing Solution, 100mL |
|
Subculture Reagent Kit | |
100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction