missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacteriophage Single-stranded DNA-binding protein gp2.5 Protein

A DNA sequence encoding the Enterobacteria phage T7 (Bacteriophage T7) Single-stranded DNA-binding protein gp2.5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9695-ye-100ug

 View more versions of this product

Product Code. 16028591

  • 1195.00 EUR / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_041970.1
Single-stranded DNA-binding protein gp2.5 Protein
Research Use Only
2.5
Recombinant Protein
Yeast
MAKKIFTSAL GTAEPYAYIA KPDYGNEERG FGNPRGVYKV DLTIPNKDPR CQRMVDEIVK CHEEAYAAAV EEYEANPPAV ARGKKPLKPY EGDMPFFDNG DGTTTFKFKC YASFQDKKTK ETKHINLVVV DSKGKKMEDV PIIGGGSKLK VKYSLVPYKW NTAVGASVKL QLESVMLVEL ATFGGGEDDW ADEVEENGYV ASGSAKASKP RDEESWDEDD EESEEADEDG DF
Greater than 90% as determined by SDS-PAGE.
1261080
100 μg
Not determined.
Full Length
Bacteriophage
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers