missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis A HAV P2C-P3B Protein
A cDNA sequence encoding the HAV P2C-P3B was constructed and used to recombinantly synthesize the protein.
208.00 EUR - 1595.00 EUR
Specifications
Name | Hepatitis A HAV P2C-P3B Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis A |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15939398
|
enQuireBio™
QP12109-100ug |
100 μg |
208.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15959398
|
enQuireBio™
QP12109-500ug |
500 μg |
795.00 EUR
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15949398
|
enQuireBio™
QP12109-1mg |
1 mg |
1595.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Hepatitis A HAV P2C-P3B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis A | |
GST | |
HAV P2C-P3B protein is >90% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10mM Tris-HCl, pH 9.6, 0.1% SDS and 50% glycerol. |