missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBsAg adw Protein
A cDNA sequence encoding the HBsAg adw was constructed and used to recombinantly synthesize the protein.
120.00 EUR - 1290.00 EUR
Specifications
Name | Hepatitis B HBsAg adw Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis B |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969448
|
enQuireBio™
QP12125-20ug |
20 μg |
120.00 EUR
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15949448
|
enQuireBio™
QP12125-100ug |
100 μg |
211.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15959448
|
enQuireBio™
QP12125-1mg |
1 mg |
1290.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Hepatitis B HBsAg adw Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE and HPLC. |
Research Use Only | |
Recombinant Protein | |
P. pastoris | |
Sterile Filtered solution containing 20mM Phosphate Buffer, 154mM sodium chloride, pH 7.1. |