missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBV core delta Protein
A cDNA sequence encoding the HBV core delta was constructed and used to recombinantly synthesize the protein.
208.00 EUR - 1595.00 EUR
Specifications
Name | Hepatitis B HBV core delta Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis B |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959468
|
enQuireBio™
QP12131-100ug |
100 μg |
208.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15979468
|
enQuireBio™
QP12131-500ug |
500 μg |
795.00 EUR
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15969468
|
enQuireBio™
QP12131-1mg |
1 mg |
1595.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Hepatitis B HBV core delta Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
Untagged | |
HBV Core Delta protein is >95% pure as determined by SDS-PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
7.5mM Phosphate buffer pH 7.2, 75mM NaCl, and 50% glycerol. |