missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV Combined Protein
A cDNA sequence encoding the HCV Combined was constructed and used to recombinantly synthesize the protein.
248.00 EUR - 1575.00 EUR
Specifications
Name | Hepatitis C HCV Combined Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15939488
|
enQuireBio™
QP12137-100ug |
100 μg |
248.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15959488
|
enQuireBio™
QP12137-500ug |
500 μg |
943.00 EUR
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15949488
|
enQuireBio™
QP12137-1mg |
1 mg |
1575.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Hepatitis C HCV Combined Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged | |
HCV protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Na-PO4, pH 8.5, 2.4mM EDTA, 5mM DTT and 0.1% SDS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title