missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 gag p17-p24 Protein
A cDNA sequence encoding the HIV-1 gag p17-p24 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12243-500ug
This item is not returnable.
View return policy
Specifications
HIV-1 gag p17-p24 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM PBS pH 7.8, 20mM NaCl 0.5M, 1mM DTT and 8M urea. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction