missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 Protein
A cDNA sequence encoding the HIV-1 was constructed and used to recombinantly synthesize the protein.
208.00 EUR - 1330.00 EUR
Specifications
Name | HIV-1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HIV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959798
|
enQuireBio™
QP12240-100ug |
100 μg |
208.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15979798
|
enQuireBio™
QP12240-500ug |
500 μg |
795.00 EUR
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15969798
|
enQuireBio™
QP12240-1mg |
1 mg |
1330.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
HIV-1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
100mM NaPO4, pH 6 and 0.05% SDS. |