missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HSV HCV Core 169aa Protein

A cDNA sequence encoding the HCV Core 169aa was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12141-500ug

 View more versions of this product

Product Code. 15979498

  • 943.00 EUR / 500µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

HSV HCV Core 169aa Protein
Research Use Only
Recombinant Protein
E. coli
Sterile filtered solution containing Phosphate saline buffer, 50mM arginine and 1M urea.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HSV
His
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.