missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Alpha-crystallin B chain Protein

A DNA sequence encoding the Homo sapiens (Human) Alpha-crystallin B chain, was expressed in the hosts and tags indicated.

160.00 EUR - 2090.00 EUR

Specifications

Accession Number NP_001276736.1
Gene ID (Entrez) 1410
Name Alpha-crystallin B chain Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15968339
View Documents Promotion Details
enQuireBio™
QP5870-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15908349
View Documents Promotion Details
enQuireBio™
QP5870-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15958339
View Documents Promotion Details
enQuireBio™
QP5870-ec-100ug
100 μg
463.00 EUR
100µg
Please to purchase this item. Need a web account? Register with us today!
15988339
View Documents Promotion Details
enQuireBio™
QP5870-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15998339
View Documents Promotion Details
enQuireBio™
QP5870-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15978339
View Documents Promotion Details
enQuireBio™
QP5870-ec-1mg
1 mg
2090.00 EUR
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001276736.1
Alpha-crystallin B chain Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
1410
Research Use Only
CRYAB
Recombinant Protein
E. coli
MDIAIHHPWI RRPFFPFHSP SRLFDQFFGE HLLESDLFPT STSLSPFYLR PPSFLRAPSW FDTGLSEMRL EKDRFSVNLD VKHFSPEELK VKVLGDVIEV HGKHEERQDE HGFISREFHR KYRIPADVDP LTITSSLSSD GVLTVNGPRK QVSGPERTIP ITREEKPAVT AAPKK
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications