missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human BJHCC20A GST (N-Term) Protein
Recombinant Protein
616.61 EUR - 784.00 EUR
Specifications
For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
---|---|
Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
Gene ID (Entrez) | 54967 |
Molecular Weight (g/mol) | 55.5 kDa |
Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18331349
|
Novus Biologicals™
H00054967-P01-25ug |
25 μg |
784.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18480310
|
Novus Biologicals™
H00054967-P01-10ug |
10 μg |
616.61 EUR
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.Specifications
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
54967 | |
Store at -80°C. Avoid freeze-thaw cycles. | |
CXORF48 | |
BJHCC20A |
50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
55.5 kDa | |
BJHCC20A, Cancer/testis antigen 55, chromosome X open reading frame 48, CT55, CT55 BRCA2-interacting protein, CXorf48, FLJ20527, RP13-565O16.1, Tumor antigen BJ-HCC-20 | |
>80% by SDS-PAGE and Coomassie blue staining |