missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CD35 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand: R&D Systems™ 5748-CD-050
329.15 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human CD35 Protein is derived from NS0. The Recombinant Human CD35 Protein has been validated for the following applications: Binding Activity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
213 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
1378 | |
CD35 | |
Mouse myeloma cell line,NS0-derived human CD35 protein Gln42-Asp1971,with a C-terminal 10-His tag and a substitution of His1208Arg | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |