missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRADD / RAIDD Protein
A DNA sequence encoding the Homo sapiens (Human) CRADD / RAIDD, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5865-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001307028.1 | |
CRADD / RAIDD Protein | |
Research Use Only | |
CRADD | |
Recombinant Protein | |
E. coli | |
MEARDKQVLR SLRLELGAEV LVEGLVLQYL YQEGILTENH IQEINAQTTG LRKTMLLLDI LPSRGPKAFD TFLDSLQEFP WVREKLKKAR EEAMTDLPAG DRLTGIPSHI LNSSPSDRQI NQLAQRLGPE WEPMVLSLGL SQTDIYRCKA NHPHNVQSQV VEAFIRWRQR FGKQATFQSL HNGLRAVEVD PSLLLHMLE | |
Greater than 90% as determined by SDS-PAGE. |
8738 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |