missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Estrogen Receptor 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Estrogen Receptor 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6000-ye-1mg
This item is not returnable.
View return policy
Specifications
NP_001035365.1 | |
Estrogen Receptor 2 Protein | |
Research Use Only | |
ESR2 | |
Recombinant Protein | |
Yeast | |
DIKNSPSSLN SPSSYNCSQS ILPLEHGSIY IPSSYVDSHH EYPAMTFYSP AVMNYSIPSN VTNLEGGPGR QTTSPNVLWP TPGHLSPLVV HRQLSHLYAE PQKSPWCEAR SLEHTLPVNR ETLKRKVSGN RCASPVTGPG SKRDAHFCAV CSDYASGYHY GVWSCEGCKA FFKRSIQGHN DYICPATNQC TIDKNRRKSC QACRLRKCYE VGMVKCGSRR ERCGYRLVRR QRSADEQLHC AGKAKRSGGH APRVRELLLD ALSPEQLVLT LLEAEPPHVL ISRPSAPFTE ASMMMSLTKL ADKELVHMIS WAKKIPGFVE LSLFDQVRLL ESCWMEVLMM GLMWRSIDHP GKLIFAPDLV LDRDEGKCVE GILEIFDMLL ATTSRFRELK LQHKEYLCVK AMILLNSSMY PLVTATQDAD SSRKLAHLLN AVTDALVWVI AKSGISSQQQ SMRLANLLML LSHVRHASNK GMEHLLNMKC KNVVPVYDLL LEMLNAHVLR GCKSSITGSE CSPAEDSKSK EGSQNPQSQ | |
Greater than 90% as determined by SDS-PAGE. |
2100 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction