missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ETHE1 / HSCO Protein
A DNA sequence encoding the Homo sapiens (Human) ETHE1 / HSCO, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6004-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_001307796.1 | |
ETHE1 / HSCO Protein | |
Research Use Only | |
ETHE1 | |
Recombinant Protein | |
E. coli | |
VARRQLSQRG GSGAPILLRQ MFEPVSCTFT YLLGDRESRE AVLIDPVLET APRDAQLIKE LGLRLLYAVN THCHADHITG SGLLRSLLPG CQSVISRLSG AQADLHIEDG DSIRFGRFAL ETRASPGHTP GCVTFVLNDH SMAFTGDALL IRGCGRTDFQ QGCAKTLYHS VHEKIFTLPG DCLIYPAHDY HGFTVSTVEE ERTLNPRLTL SCEEFVKIMG NLNLPKPQQI DFAVPANMRC GVQTPTA | |
Greater than 90% as determined by SDS-PAGE. |
23474 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction