missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FABP1 Protein
A cDNA sequence encoding the FABP1 was constructed and used to recombinantly synthesize the protein.
143.00 EUR - 4410.00 EUR
Specifications
Name | FABP1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15958518
|
enQuireBio™
QP11832-5ug |
5 μg |
143.00 EUR
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15948518
|
enQuireBio™
QP11832-25ug |
25 μg |
222.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15938518
|
enQuireBio™
QP11832-1mg |
1 mg |
4410.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
FABP1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein solution (1 mg/ml) contains 20mM Tris-HCl pH 8. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSFSGKYQLQ SQENFEAFMK AIGLPEELIQ KGKDIKGVSEIVQNGKHFKF TITAGSKVIQ NEFTVGEECE LETMTGEKVK TVVQLEGDNK LVTTFKNIKSVTELNGDIIT NTMTLGDIVF KRISKRI | |
Greater than 95.0% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title