missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FAM3D Protein

A cDNA sequence encoding the FAM3D was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11847-2ug

 View more versions of this product

Product Code. 15978568

  • 143.00 EUR / 2µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

FAM3D Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASYMSFSMKTIR LPRWLAASPT KEIQVKKYKC GLIKPCPANY FAFKICSGAA NVVGPTMCFE DRMIMSPVKN NVGRGLNIAL VNGTTGAVLG QKAFDMYSGD VMHLVKFLKE IPGGALVLVA SYDDPGTKMN DESRKLFSDL GSSYAKQLGF RDSWVFIGAK DLRGKSPFEQ FLKNSPDTNK YEGWPELLEM EGCMPPKPF
Greater than 90.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
FAM3D filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4.0.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers