missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FDPS Protein
A DNA sequence encoding the Homo sapiens (Human) FDPS, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6032-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_001129293.1 | |
FDPS Protein | |
Research Use Only | |
FDPS | |
Recombinant Protein | |
E. coli | |
MPLSRWLRSV GVFLLPAPYW APRERWLGSL RRPSLVHGYP VLAWHSARCW CQAWTEEPRA LCSSLRMNGD QNSDVYAQEK QDFVQHFSQI VRVLTEDEMG HPEIGDAIAR LKEVLEYNAI GGKYNRGLTV VVAFRELVEP RKQDADSLQR AWTVGWCVEL LQAFFLVADD IMDSSLTRRG QICWYQKPGV GLDAINDANL LEACIYRLLK LYCREQPYYL NLIELFLQSS YQTEIGQTLD LLTAPQGNVD LVRFTEKRYK SIVKYKTAFY SFYLPIAAAM YMAGIDGEKE HANAKKILLE MGEFFQIQDD YLDLFGDPSV TGKIGTDIQD NKCSWLVVQC LQRATPEQYQ ILKENYGQKE AEKVARVKAL YEELDLPAVF LQYEEDSYSH IMALIEQYAA PLPPAVFLGL ARKIYKRRK | |
Greater than 90% as determined by SDS-PAGE. |
2224 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |