missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human G-CSF / CSF3 Protein
A DNA sequence encoding the Homo sapiens (Human) G-CSF / CSF3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8545-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_000750.1 | |
G-CSF / CSF3 Protein | |
Research Use Only | |
CSF3 | |
Recombinant Protein | |
E. coli | |
VQEATPLGPA SSLPQSFLLK CLEQVRKIQG DGAALQEKLV SECATYKLCH PEELVLLGHS LGIPWAPLSS CPSQALQLAG CLSQLHSGLF LYQGLLQALE GISPELGPTL DTLQLDVADF ATTIWQQMEE LGMAPALQPT QGAMPAFASA FQRRAGGVLV ASHLQSFLEV SYRV | |
Greater than 90% as determined by SDS-PAGE. |
1440 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |