missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GCHFR Protein
A cDNA sequence encoding the GCHFR was constructed and used to recombinantly synthesize the protein.
120.00 EUR - 3585.00 EUR
Specifications
Name | GCHFR Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15968868
|
enQuireBio™
QP11940-5ug |
5 μg |
120.00 EUR
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15958868
|
enQuireBio™
QP11940-20ug |
20 μg |
186.00 EUR
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15948868
|
enQuireBio™
QP11940-1mg |
1 mg |
3585.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
GCHFR Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GCHFR protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 40% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPYLLIS TQIRMEVGPT MVGDEQSDPE LMQHLGASKR RALGNNFYEY YVDDPPRIVL DKLERRGFRV LSMTGVGQTL VWCLHKE | |
Greater than 95.0% as determined by SDS-PAGE. |