missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GLRX5 Protein
A cDNA sequence encoding the GLRX5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11992-25ug
This item is not returnable.
View return policy
Specifications
51218 | |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSGSLGRAAA ALLRWGRGAG GGGLWGPGVR AAGSGAGGGG SAEQLDALVK KDKVVVFLKG TPEQPQCGFS NAVVQILRLH GVRDYAAYNV LDDPELRQGI KDYSNWPTIP QVYLNGEFVG GCDILLQMHQ NGDLVEELKK LGIHSALLDE KKDQDSK | |
Greater than 85% as determined by SDS-PAGE. |
GLRX5 Protein | |
Research Use Only | |
GLRX5 | |
Human | |
His | |
GLRX5 solution containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 0.1M NaCl. |