missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GMFB His Protein
A cDNA sequence encoding the GMFB His was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12005-5ug
This item is not returnable.
View return policy
Specifications
GMFB His Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSESLVVCDV AEDLVEKLRK FRFRKETNNA AIIMKIDKDK RLVVLDEELE GISPDELKDE LPERQPRFIV YSYKYQHDDG RVSYPLCFIF SSPVGCKPEQ QMMYAGSKNK LVQTAELTKV FEIRNTEDLT EEWLREKLGF FH | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GMFB 1 mg/ml protein solution contains 20mM Tris-HCL pH 8.0, 1mM DTT, 0.1M NaCl and 10% Glycerol. |