missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human GPT2 His Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP3-18182-250UG
This item is not returnable.
View return policy
Description
Specific activity is > 100units/mg, and is defined as the amount of enzyme that cleaves 1 umole of L-Alanine to L-Glutamate per minute at pH 7.5 at 37CSpecifications
0.5 mg/ml | |
20mM Tris-HCl buffer (pH 7.5), 30% glycerol, 2mM DTT, 0.2M NaCl | |
M.W. Theoretical: 60.3 kDa | |
Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles. | |
GPT2 | |
>90%, by SDS-PAGE |
Bioactivity, SDS-PAGE | |
84706 | |
250 μg | |
AAT2, alanine aminotransferase 2, ALT2EC 2.6.1.2, Glutamate pyruvate transaminase 2, glutamic pyruvate transaminase (alanine aminotransferase) 2, Glutamic--alanine transaminase 2, glutamic-pyruvate transaminase 2, Glutamic--pyruvic transaminase 2, GPT 2 | |
metabolism | |
GPT2 |