missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Hemopexin Protein

A cDNA sequence encoding the Hemopexin was constructed and used to recombinantly synthesize the protein.

143.00 EUR - 2525.00 EUR

Specifications

Name Hemopexin Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15949718
View Documents
enQuireBio™
QP12215-10ug
10 μg
143.00 EUR
10µg
Please to purchase this item. Need a web account? Register with us today!
15969718
View Documents
enQuireBio™
QP12215-50ug
50 μg
222.00 EUR
50µg
Please to purchase this item. Need a web account? Register with us today!
15959718
View Documents
enQuireBio™
QP12215-1mg
1 mg
2525.00 EUR
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Hemopexin Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Lyophilized from (1.22 mg/ml) solution containing 11.9mM phosphate buffer, 137mM NaCl and 2.7mM KCl, pH 7.4.
Research Use Only
Recombinant Protein
Human
MARVLGAPVA LGLWSLCWSL AIATPLPPTS AHGNVAEGET KPDPDVTERC SDGWSFDATT LDDNGTMLFF KGEFVWKSHK WDRELISERW KNFPSPVDAA FRQGHNSVFL IKGDKVWVYP PEKKEKGYPK LLQDEFPGIP SPLDAAVECH RGECQAEGVL FFQGDREWFW DLATGTMKER SWPAVGNCSS ALRWLGRYYC FQGNQFLRFD PVRGEVPPRY PRDVRDYFMP CPGRGHGHRN GTGHGNSTHH GPEYMRCSPH LVLSALTSDN HGATYAFSGT HYWRLDTSRD GWHSWPIAHQ WPQGPSAVDA AFSWEEKLYL VQGTQVYVFL TKGGYTLVSG YPKRLEKEVG TPHGIILDSV DAAFICPGSS RLHIMAGRRL WWLDLKSGAQ ATWTELPWPH EKVDGALCME KSLGPNSCSA NGPGLYLIHG PNLYCYSDVE KLNAAKALPQ PQNVTSLLGC TH
Greater than 95.0%.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.