missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HNMT Protein

A cDNA sequence encoding the HNMT was constructed and used to recombinantly synthesize the protein.

120.00 EUR - 4780.00 EUR

Specifications

Name HNMT Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15979948
View Documents Promotion Details
enQuireBio™
QP12289-5ug
5 μg
120.00 EUR
5µg
Please to purchase this item. Need a web account? Register with us today!
15969948
View Documents Promotion Details
enQuireBio™
QP12289-20ug
20 μg
186.00 EUR
20µg
Please to purchase this item. Need a web account? Register with us today!
15959948
View Documents Promotion Details
enQuireBio™
QP12289-1mg
1 mg
4780.00 EUR
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

HNMT Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The HNMT solution contains 20mM Tris-HCl pH 8 and 10% glycerol.
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMASS MRSLFSDHGK YVESFRRFLN HSTEHQCMQE FMDKKLPGII GRIGDTKSEI KILSIGGGAG EIDLQILSKV QAQYPGVCIN NEVVEPSAEQ IAKYKELVAK TSNLENVKFA WHKETSSEYQ SRMLEKKELQ KWDFIHMIQM LYYVKDIPAT LKFFHSLLGT NAKMLIIVVS GSSGWDKLWK KYGSRFPQDD LCQYITSDDL TQMLDNLGLK YECYDLLSTM DISDCFIDGD ENGDLLWDFL TETCNFNATA PPDLRAELGK DLQEPEFSAK KEGKVLFNNT LSFIVIEA
Greater than 95.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications